Learn More
Medchemexpress LLC Omeprazole metabolite omeprazole sulfone (methoxy-d3) | 1189891-71-1 | 99.7% | C17H16D3N3O4S | 1 MG
SDP
Catalog No. 5000432717
Encompass_Preferred
Omeprazole metabolite omeprazole sulfone (methoxy-d3) is a deuterium labeled metabolite of omeprazole, a proton pump inhibitor. This compound serves multiple purposes in research and development.
  • It can be utilized as a tracer in various studies.
  • It serves as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily for quantitation during the drug development process.
  • Deuteration has shown potential to influence the pharmacokinetic and metabolic profiles of pharmaceutical compounds.
Catalog No. 50-004-32717 Supplier Medchemexpress LLC Supplier No. HYB0113S21MG
Omeprazole metabolite omeprazole sulfone (methoxy-d3) is a deuterium labeled metabolite of omeprazole, a proton pump inhibitor. This compound serves multiple purposes in research and development.
  • It can be utilized as a tracer in various studies.
  • It serves as an internal standard for quantitative analysis through methods like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily for quantitation during the drug development process.
  • Deuteration has shown potential to influence the pharmacokinetic and metabolic profiles of pharmaceutical compounds.