Learn More
Cayman Chemical Omeprazole N-oxide, Purity: ≥98%, CAS: 176219-04-8, 10 mg
SDP
Catalog No. 502913950
Encompass_Preferred
A metabolite of omeprazole; a potential impurity found in commercial preparations of omeprazole
Catalog No. 50-291-3950 Supplier Cayman Chemical Supplier No. 3543010 MG
A metabolite of omeprazole; a potential impurity found in commercial preparations of omeprazole