Learn More
Cayman Chemical Omeprazole N-oxide | 176219-04-8 | 25mg
SDP
Catalog No. 502909587
Encompass_Preferred
A metabolite of omeprazole; a potential impurity found in commercial preparations of omeprazole
Catalog No. 50-290-9587 Supplier Cayman Chemical Supplier No. 3543025 MG
A metabolite of omeprazole; a potential impurity found in commercial preparations of omeprazole