Learn More
Medchemexpress LLC Omeprazole sodium | 95510-70-6 | 99.1% | 100 MG
SDP
Catalog No. 5000432885
Encompass_Preferred
Omeprazole sodium is a proton pump inhibitor (PPI) used for treating acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity with a Ki of 2 to 6 μM. It also inhibits the growth of Gram-positive and Gram-negative bacteria and acts as a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).
  • Used for acid-related gastrointestinal disorders
  • Competitively inhibits CYP2C19 activity with a Ki of 2 to 6 μM
  • Inhibits the growth of Gram-positive and Gram-negative bacteria
  • Potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor)
  • Used in the study of dyspepsia, peptic ulcer disease, gastroesophageal reflux disease, laryngopharyngeal reflux, and Zollinger-Ellison syndrome.
Catalog No. 50-004-32885 Supplier Medchemexpress LLC Supplier No. HYB0113A100MG
Omeprazole sodium is a proton pump inhibitor (PPI) used for treating acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity with a Ki of 2 to 6 μM. It also inhibits the growth of Gram-positive and Gram-negative bacteria and acts as a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).
  • Used for acid-related gastrointestinal disorders
  • Competitively inhibits CYP2C19 activity with a Ki of 2 to 6 μM
  • Inhibits the growth of Gram-positive and Gram-negative bacteria
  • Potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor)
  • Used in the study of dyspepsia, peptic ulcer disease, gastroesophageal reflux disease, laryngopharyngeal reflux, and Zollinger-Ellison syndrome.