Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Omeprazole (sodium) | 95510-70-6 | 99.1% | C17H18N3NaO3S | 1 ML
SDP

Catalog No. 5000432431
Encompass_Preferred

Omeprazole sodium is a proton pump inhibitor (PPI) used to treat acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity and also inhibits the growth of Gram-positive and Gram-negative bacteria. It acts as a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor.

  • Proton pump inhibitor
  • Treats acid-related gastrointestinal disorders
  • Inhibits CYP2C19 activity
  • Inhibits growth of Gram-positive and Gram-negative bacteria
  • Potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor
  • Purity: 99.1%
  • Chemical formula: C17H18N3NaO3S
  • Molecular Weight: 367.40
  • Appearance: Solid
  • Melting/freezing point: >165°C

Catalog No. 50-004-32431 Supplier Medchemexpress LLC Supplier No. HYB0113A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Omeprazole sodium is a proton pump inhibitor (PPI) used to treat acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity and also inhibits the growth of Gram-positive and Gram-negative bacteria. It acts as a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor.

  • Proton pump inhibitor
  • Treats acid-related gastrointestinal disorders
  • Inhibits CYP2C19 activity
  • Inhibits growth of Gram-positive and Gram-negative bacteria
  • Potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor
  • Purity: 99.1%
  • Chemical formula: C17H18N3NaO3S
  • Molecular Weight: 367.40
  • Appearance: Solid
  • Melting/freezing point: >165°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.