Learn More
Medchemexpress LLC Omeprazole sodium | 95510-70-6 | MFCD07802816 | 99.10% | C17H18N3NaO3S | 500 MG
SDP
Catalog No. 5000432866
Encompass_Preferred
Omeprazole sodium is a proton pump inhibitor (PPI) used in the treatment of acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity with a Ki of 2 to 6 μM. It also inhibits the growth of both Gram-positive and Gram-negative bacteria, and is a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).
  • Proton pump inhibitor (PPI)
  • Treats acid-related gastrointestinal disorders
  • Competitively inhibits CYP2C19 activity with a Ki of 2 to 6 μM
  • Inhibits growth of Gram-positive and Gram-negative bacteria
  • Potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor)
  • Utilized in the study of dyspepsia, peptic ulcer disease, gastroesophageal reflux disease, laryngopharyngeal reflux, and Zollinger-Ellison syndrome
Catalog No. 50-004-32866 Supplier Medchemexpress LLC Supplier No. HYB0113A500MG
Omeprazole sodium is a proton pump inhibitor (PPI) used in the treatment of acid-related gastrointestinal disorders. It competitively inhibits CYP2C19 activity with a Ki of 2 to 6 μM. It also inhibits the growth of both Gram-positive and Gram-negative bacteria, and is a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).
  • Proton pump inhibitor (PPI)
  • Treats acid-related gastrointestinal disorders
  • Competitively inhibits CYP2C19 activity with a Ki of 2 to 6 μM
  • Inhibits growth of Gram-positive and Gram-negative bacteria
  • Potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor)
  • Utilized in the study of dyspepsia, peptic ulcer disease, gastroesophageal reflux disease, laryngopharyngeal reflux, and Zollinger-Ellison syndrome