Learn More
Selleck Chemical LLC Omeprazole Sodium
SDP
Catalog No. 5000003058
Encompass_Preferred
Omeprazole Sodium is the sodium salt form of omeprazole which is a proton pump inhibitor and suppresses gastric acid secretion Omeprazole Sodium is a metabolism-dependent inhibitor (MDI) of CYP2C19
Catalog No. 50-000-03058 Supplier Selleck Chemical LLC Supplier No. S565825MG
Omeprazole Sodium is the sodium salt form of omeprazole which is a proton pump inhibitor and suppresses gastric acid secretion Omeprazole Sodium is a metabolism-dependent inhibitor (MDI) of CYP2C19