Learn More
Medchemexpress LLC Omeprazole (Standard) | 73590-58-6 | 99.9% | C17H19N3O3S | 5 MG
SDP
Catalog No. 5000432676
Encompass_Preferred
Omeprazole (Standard) is an analytical standard used for research and analytical applications, such as qualitative, quantitative, and methodological experiments in HPLC, GC, and MS.
  • Analytical standard of omeprazole
  • Proton pump inhibitor (PPI)
  • Competitive inhibitor of CYP2C19 activity
  • Inhibits growth of Gram-positive and Gram-negative bacteria
  • Potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor
Catalog No. 50-004-32676 Supplier Medchemexpress LLC Supplier No. HYB0113R5MG
Omeprazole (Standard) is an analytical standard used for research and analytical applications, such as qualitative, quantitative, and methodological experiments in HPLC, GC, and MS.
  • Analytical standard of omeprazole
  • Proton pump inhibitor (PPI)
  • Competitive inhibitor of CYP2C19 activity
  • Inhibits growth of Gram-positive and Gram-negative bacteria
  • Potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor