Learn More
Medchemexpress LLC Omeprazole (Standard) | 73590-58-6 | 99.9% | C17H19N3O3S | 50 MG
SDP
Catalog No. 5000432675
Encompass_Preferred
Omeprazole (Standard) is an analytical reference standard and a proton pump inhibitor (PPI) used for acid-related gastrointestinal disorders. This product is designed for research, inhibiting CYP2C19 activity (Ki 2-6 μM) and the growth of Gram-positive and Gram-negative bacteria. It also acts as a potent brain-penetrant neutral sphingomyelinase (N-SMase) and exosome inhibitor.
  • Analytical standard for qualitative, quantitative, and methodological research experiments (HPLC, GC, MS).
  • Treats acid-related gastrointestinal disorders.
  • Competitively inhibits CYP2C19 activity (Ki 2-6 μM).
  • Inhibits Gram-positive and Gram-negative bacteria growth.
  • Potent brain-penetrant neutral sphingomyelinase (N-SMase) and exosome inhibitor.
  • High purity of 99.86%.
  • Solid, white to off-white appearance.
Catalog No. 50-004-32675 Supplier Medchemexpress LLC Supplier No. HYB0113R50MG
Omeprazole (Standard) is an analytical reference standard and a proton pump inhibitor (PPI) used for acid-related gastrointestinal disorders. This product is designed for research, inhibiting CYP2C19 activity (Ki 2-6 μM) and the growth of Gram-positive and Gram-negative bacteria. It also acts as a potent brain-penetrant neutral sphingomyelinase (N-SMase) and exosome inhibitor.
  • Analytical standard for qualitative, quantitative, and methodological research experiments (HPLC, GC, MS).
  • Treats acid-related gastrointestinal disorders.
  • Competitively inhibits CYP2C19 activity (Ki 2-6 μM).
  • Inhibits Gram-positive and Gram-negative bacteria growth.
  • Potent brain-penetrant neutral sphingomyelinase (N-SMase) and exosome inhibitor.
  • High purity of 99.86%.
  • Solid, white to off-white appearance.