Learn More
Cayman Chemical Omeprazole sulfide, Purity: ≥98%, CAS: 73590-85-9, 250 mg
SDP
Catalog No. 502984536
Encompass_Preferred
An active metabolite of omeprazole; has been used as a precursor in the synthesis of esomeprazole; a potential impurity in commercial preparations of omeprazole
Catalog No. 50-298-4536 Supplier Cayman Chemical Supplier No. 18885250 MG
An active metabolite of omeprazole; has been used as a precursor in the synthesis of esomeprazole; a potential impurity in commercial preparations of omeprazole