Learn More
Cayman Chemical Omeprazole sulfIde 500mg
SDP
Catalog No. 502984754
Encompass_Preferred
An active metabolite of omeprazole; has been used as a precursor in the synthesis of esomeprazole; a potential impurity in commercial preparations of omeprazole
Catalog No. 50-298-4754 Supplier Cayman Chemical Supplier No. 18885500 MG
An active metabolite of omeprazole; has been used as a precursor in the synthesis of esomeprazole; a potential impurity in commercial preparations of omeprazole