Learn More
Cayman Chemical Omeprazole sulfIde-d3 5mg
SDP
Catalog No. 502945797
Encompass_Preferred
An internal standard for the quantification of omeprazole sulfide by GC- or LC-MS
Catalog No. 50-294-5797 Supplier Cayman Chemical Supplier No. 402605 MG
An internal standard for the quantification of omeprazole sulfide by GC- or LC-MS