Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Omeprazole sulfide-d3 | 922730-98-1 | 99.8% | 332.44 | C17H16D3N3O2S | 5 MG
SDP

Catalog No. 5000221698
Encompass_Preferred

Omeprazole sulfide-d3 is the deuterium-labeled analogue of omeprazole sulfide, a metabolite of the proton-pump inhibitor omeprazole. It is provided as an isotopically labeled reference standard intended for use as a tracer and an internal standard in quantitative analytical workflows to support accurate identification and measurement.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High purity (99.79% by LCMS) for reliable assay performance.
  • Molecular weight 332.44 and formula C17H16D3N3O2S for unambiguous identification.
  • Available in solid and solution formats for flexible use.

Catalog No. 50-002-21698 Supplier Medchemexpress LLC Supplier No. HY141776S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Omeprazole sulfide-d3 is the deuterium-labeled analogue of omeprazole sulfide, a metabolite of the proton-pump inhibitor omeprazole. It is provided as an isotopically labeled reference standard intended for use as a tracer and an internal standard in quantitative analytical workflows to support accurate identification and measurement.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High purity (99.79% by LCMS) for reliable assay performance.
  • Molecular weight 332.44 and formula C17H16D3N3O2S for unambiguous identification.
  • Available in solid and solution formats for flexible use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.