Learn More
Medchemexpress LLC Omeprazole sulfide-d3 | 922730-98-1 | 99.8% | C17H16D3N3O2S | 1 ML
SDP
Catalog No. 5000356539
Encompass_Preferred
Omeprazole sulfide-d3 is the deuterium labeled form of Omeprazole sulfide, a metabolite of the proton pump inhibitor Omeprazole. It serves as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration can impact drug pharmacokinetic and metabolic profiles.
  • Deuterium labeled omeprazole sulfide
  • Metabolite of omeprazole, a proton pump inhibitor
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
  • Prepare working solutions fresh for in vivo experiments
  • Heat or sonicate for dissolution if precipitation occurs
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-56539 Supplier Medchemexpress LLC Supplier No. HY141776S10MM/1ML
Omeprazole sulfide-d3 is the deuterium labeled form of Omeprazole sulfide, a metabolite of the proton pump inhibitor Omeprazole. It serves as a tracer and internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration can impact drug pharmacokinetic and metabolic profiles.
  • Deuterium labeled omeprazole sulfide
  • Metabolite of omeprazole, a proton pump inhibitor
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
  • Prepare working solutions fresh for in vivo experiments
  • Heat or sonicate for dissolution if precipitation occurs
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month