Learn More
Medchemexpress LLC Omeprazole sulfide-d3 | 922730-98-1 | MFCD09841123 | 99.8% | C17H16D3N3O2S | 10MG
SDP
Catalog No. 5000261665
Encompass_Preferred
Omeprazole sulfide-d3 is the deuterium-labeled analogue of omeprazole sulfide, supplied as a stable isotope reference standard for analytical and metabolic studies. It is intended for use as an internal standard in quantitative mass spectrometry and pharmacokinetic investigations.
  • Deuterium-labeled internal standard for LC-MS quantitation.
  • High purity (99.8%).
  • Suitable for metabolic and pharmacokinetic studies.
  • Small package sizes convenient for analytical workflows.
  • Stable as a powder at -20 °C for long-term storage; follow solvent storage recommendations.
  • Handling notes: hygroscopic DMSO affects solubility; aliquot and avoid repeated freeze-thaw.
Catalog No. 50-002-61665 Supplier Medchemexpress LLC Supplier No. HY141776S10MG
Omeprazole sulfide-d3 is the deuterium-labeled analogue of omeprazole sulfide, supplied as a stable isotope reference standard for analytical and metabolic studies. It is intended for use as an internal standard in quantitative mass spectrometry and pharmacokinetic investigations.
  • Deuterium-labeled internal standard for LC-MS quantitation.
  • High purity (99.8%).
  • Suitable for metabolic and pharmacokinetic studies.
  • Small package sizes convenient for analytical workflows.
  • Stable as a powder at -20 °C for long-term storage; follow solvent storage recommendations.
  • Handling notes: hygroscopic DMSO affects solubility; aliquot and avoid repeated freeze-thaw.