Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Omeprazole Sulfide 98.0+%, TCI America™
SDP

Catalog No. O0437200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0437200MG 200 mg
O04371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0437200MG Supplier TCI America Supplier No. O0437200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73590-85-9
Molecular Formula C17H19N3O2S
Molecular Weight (g/mol) 329.418
MDL Number MFCD00869021
InChI Key XURCIPRUUASYLR-UHFFFAOYSA-N
Synonym 5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridyl)methyl]thio]benzimidazole
PubChem CID 155794
IUPAC Name 6-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfanyl]-1H-benzimidazole
SMILES CC1=CN=C(C(=C1OC)C)CSC2=NC3=C(N2)C=C(C=C3)OC

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 200 mg
Formula Weight 329.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Omeprazole Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.