Learn More
Cayman Chemical Omeprazole sulfon-d3 500ug
SDP
Catalog No. 502936727
Encompass_Preferred
An internal standard for the quantification of omeprazole sulfone by GC- or LC-MS
Catalog No. 50-293-6727 Supplier Cayman Chemical Supplier No. 26784500 UG
An internal standard for the quantification of omeprazole sulfone by GC- or LC-MS