Learn More
Apexbio Technology LLC OMEPRAZOLE-SULFONE-10MG
SDP
Catalog No. 5000569399
Encompass_Preferred
Omeprazole sulfone (CAS No 88546-55-8) is a key metabolite of omeprazole in the proton pump inhibitor category Originating from the necessity to understand omeprazole s pharmacokinetics it primarily targets the H /K ATPase enzyme system in gastric parietal cells inhibiting acid secretion Its mechanism involves the binding and alteration of specific pathways related to gastric acid production Omeprazole sulfone exhibits in vitro activity typically in the nanomolar to low micromolar range depending on experimental conditions It is commonly used in a variety of applications such as cell models animal studies and drug screening to explore gastric acid-related processes Experimental concentrations typically depend on the specific research design This compound is instrumental in evaluating the efficacy and safety of proton pump inhibitors and is pivotal in the research of gastro-related disorders and their treatment pathways
Catalog No. 50-005-69399 Supplier Apexbio Technology LLC Supplier No. C347110
Omeprazole sulfone (CAS No 88546-55-8) is a key metabolite of omeprazole in the proton pump inhibitor category Originating from the necessity to understand omeprazole s pharmacokinetics it primarily targets the H /K ATPase enzyme system in gastric parietal cells inhibiting acid secretion Its mechanism involves the binding and alteration of specific pathways related to gastric acid production Omeprazole sulfone exhibits in vitro activity typically in the nanomolar to low micromolar range depending on experimental conditions It is commonly used in a variety of applications such as cell models animal studies and drug screening to explore gastric acid-related processes Experimental concentrations typically depend on the specific research design This compound is instrumental in evaluating the efficacy and safety of proton pump inhibitors and is pivotal in the research of gastro-related disorders and their treatment pathways