Learn More
Medchemexpress LLC Omeprazole sulfone | 88546-55-8 | 99.23% | 361.42 | 5 MG
SDP
Catalog No. 5000429901
Encompass_Preferred
Omeprazole sulfone is one of the major metabolites of Omeprazole (HY-B0113). It is generated via sulfoxidation mediated by cytochrome P450 3A4 (CYP3A4).
  • For research use only.
  • Not sold to patients.
Catalog No. 50-004-29901 Supplier Medchemexpress LLC Supplier No. HYG00075MG
Omeprazole sulfone is one of the major metabolites of Omeprazole (HY-B0113). It is generated via sulfoxidation mediated by cytochrome P450 3A4 (CYP3A4).
  • For research use only.
  • Not sold to patients.