Learn More
Medchemexpress LLC Omeprazole sulfone | 88546-55-8 | MFCD00869772 | 99.2% | 361.42 g/mol | C17H19N3O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HYG000710MG
Encompass_Preferred
Omeprazole sulfone is an analytical standard and a primary metabolite of the proton-pump inhibitor omeprazole. It is formed via sulfoxidation mediated by cytochrome P450 3A4 (CYP3A4) and is provided as a high-purity reference material for metabolic, pharmacokinetic, and analytical studies.
  • Primary metabolite of omeprazole formed by CYP3A4.
  • High purity reference standard (99.23%).
  • Molecular formula C17H19N3O4S; molecular weight 361.42 g/mol.
  • Suitable for metabolic profiling and analytical method validation.
  • Available as a 10 mg vial for analytical use.
Catalog No. 50-002-97162