Learn More
Electron Microscopy Sciences Applied Physics LaB6 Cathode for Omicron
SDP
Catalog No. 50949852
Encompass
Lanthanum hexaboride (LaB6) and cerium hexaboride (CeB6) cathodes are ideal for many small spot size applications such as SEM, TEM, surface analysis and metrology, and for high current applications such as microwave tubes, lithography, electron-beam welders, X-ray sources and free electron lasers. Greater brightness from low work function yielding higher currents at lower temps than tungsten.
Type: LaB6
Angle: 90°
Tips: 100 mm
Catalog No. 50-949-852 Supplier Electron Microscopy Sciences Supplier No. 80930L
Lanthanum hexaboride (LaB6) and cerium hexaboride (CeB6) cathodes are ideal for many small spot size applications such as SEM, TEM, surface analysis and metrology, and for high current applications such as microwave tubes, lithography, electron-beam welders, X-ray sources and free electron lasers. Greater brightness from low work function yielding higher currents at lower temps than tungsten.
Type: LaB6
Angle: 90°
Tips: 100 mm