Learn More
Description
- Provides a stress-free, one-step preservation protocol with minimum hands-on time
- Optimized to preserve cells for a variety of downstream transcriptomic, proteomic and multiomic applications, including RNA-seq, CITE-seq, flow cytometry and qPCR
- Protects cell viability and preserves different cell populations in your samples for up to 72 hours at 4°C
- Developed and tested across multiple sample types: PBMCs and tissue samples
- Available in 12 x 1 mL vials
Specifications
Specifications
| For Use With (Equipment) | BD Rhapsody platform, Single-Cell Analysis System |
| Product Type | Sample Preservation Buffer |
| Format | Vial |
| Quantity | 12 x 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.