Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD™ OMICS-Guard Sample Preservation Buffer

Catalog No. BDB570911 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570911 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB570911 Supplier BD™ Supplier No. 570911
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The BD™ OMICS-Guard Sample Preservation Buffer is developed and optimized to address the need for high-quality biological sample preservation over an extended period of time when samples cannot be processed at the same time or location.

  • Provides a stress-free, one-step preservation protocol with minimum hands-on time
  • Optimized to preserve cells for a variety of downstream transcriptomic, proteomic and multiomic applications, including RNA-seq, CITE-seq, flow cytometry and qPCR
  • Protects cell viability and preserves different cell populations in your samples for up to 72 hours at 4°C
  • Developed and tested across multiple sample types: PBMCs and tissue samples
  • Available in a 50 mL bottle
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) BD Rhapsody™ platform, Single-Cell Analysis System
Includes 50 mL Bottle
Quantity 50 mL
Product Type Sample Preservation Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.