Learn More
Medchemexpress LLC HY-111406 5mg Medchemexpress, Omidenepag isopropyl CAS:1187451-19-9 Purity:>98%
SDP
Catalog No. 502080791
Encompass_Preferred
Medchemexpress, HY-111406 5mg Omidenepag isopropyl CAS:1187451-19-9 Omidenepag isopropyl is a selective EP2 receptor agonist. Omidenepag isopropyl is converted to the active product Omidenepag during corneal penetration, and Omidenepag is a highly selective EP2 receptor agonist. Omidenepag isopropyl shows only weak affinity for EP1, EP2, and FP receptors. Omidenepag isopropyl is under development for the treatment of glaucoma as an intraocular pressure (IOP)-lowering drug. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0791 Supplier Medchemexpress LLC Supplier No. HY1114065MG
Medchemexpress, HY-111406 5mg Omidenepag isopropyl CAS:1187451-19-9 Omidenepag isopropyl is a selective EP2 receptor agonist. Omidenepag isopropyl is converted to the active product Omidenepag during corneal penetration, and Omidenepag is a highly selective EP2 receptor agonist. Omidenepag isopropyl shows only weak affinity for EP1, EP2, and FP receptors. Omidenepag isopropyl is under development for the treatment of glaucoma as an intraocular pressure (IOP)-lowering drug. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.