Learn More
Cayman Chemical OmIdenpag Isopropyl 10mg
SDP
Catalog No. 502944922
Encompass_Preferred
A prodrug form of omidenepag; reduces IOP in ocular normotensive and laser-induced ocular hypertensive cynomolgus monkeys when topically administered at 0.01%/eye
Catalog No. 50-294-4922 Supplier Cayman Chemical Supplier No. 4104610 MG
A prodrug form of omidenepag; reduces IOP in ocular normotensive and laser-induced ocular hypertensive cynomolgus monkeys when topically administered at 0.01%/eye