Learn More
Medchemexpress LLC Omigapil maleate | 200189-97-5 | 99.8% | 391.42 | 25 MG
SDP
Catalog No. 5000393259
Encompass_Preferred
Omigapil maleate (CGP3466B) is an orally active GAPDH nitrosylation inhibitor that abrogates Aβ1-42-induced tau acetylation, memory impairment, and locomotor dysfunction in mice. It has potential for Alzheimer's disease research and is also an apoptosis inhibitor. Additionally, it can be used for congenital muscular dystrophy (CMD) research.
  • Orally active GAPDH nitrosylation inhibitor.
  • Abrogates Aβ1-42-induced tau acetylation, memory impairment, and locomotor dysfunction in mice.
  • Potential for Alzheimer's disease research.
  • Apoptosis inhibitor.
  • Can be used for congenital muscular dystrophy (CMD) research.
  • Click chemistry reagent with an alkyne group for CuAAc reactions.
Catalog No. 50-003-93259 Supplier Medchemexpress LLC Supplier No. 25MGHY16361A
Omigapil maleate (CGP3466B) is an orally active GAPDH nitrosylation inhibitor that abrogates Aβ1-42-induced tau acetylation, memory impairment, and locomotor dysfunction in mice. It has potential for Alzheimer's disease research and is also an apoptosis inhibitor. Additionally, it can be used for congenital muscular dystrophy (CMD) research.
  • Orally active GAPDH nitrosylation inhibitor.
  • Abrogates Aβ1-42-induced tau acetylation, memory impairment, and locomotor dysfunction in mice.
  • Potential for Alzheimer's disease research.
  • Apoptosis inhibitor.
  • Can be used for congenital muscular dystrophy (CMD) research.
  • Click chemistry reagent with an alkyne group for CuAAc reactions.