Learn More
Medchemexpress LLC Omigapil maleate | 200189-97-5 | 99.8% | 391.42 | 5 MG
SDP
Catalog No. 5000364316
Encompass_Preferred
Omigapil maleate (CGP3466B) is an orally active GAPDH nitrosylation inhibitor with potential for research into Alzheimer's disease and congenital muscular dystrophy (CMD). It is also an apoptosis inhibitor and a click chemistry reagent.
  • Abrogates Aβ1-42-induced tau acetylation, memory impairment, and locomotor dysfunction in mice
  • Acts as an apoptosis inhibitor
  • Can be used for research of congenital muscular dystrophy (CMD)
  • Functions as a click chemistry reagent containing an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-003-64316 Supplier Medchemexpress LLC Supplier No. 5MGHY16361A
Omigapil maleate (CGP3466B) is an orally active GAPDH nitrosylation inhibitor with potential for research into Alzheimer's disease and congenital muscular dystrophy (CMD). It is also an apoptosis inhibitor and a click chemistry reagent.
  • Abrogates Aβ1-42-induced tau acetylation, memory impairment, and locomotor dysfunction in mice
  • Acts as an apoptosis inhibitor
  • Can be used for research of congenital muscular dystrophy (CMD)
  • Functions as a click chemistry reagent containing an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups