Learn More
Medchemexpress LLC Omigapil maleate | 200189-97-5 | 99.8% | 391.42 | 50 MG
SDP
Catalog No. 5000392572
Encompass_Preferred
Omigapil maleate (CGP3466B) is an orally active GAPDH nitrosylation inhibitor that can abrogate Aβ1-42-induced tau acetylation, memory impairment, and locomotor dysfunction in mice. It has potential for research into Alzheimer's disease and congenital muscular dystrophy (CMD). Omigapil maleate also functions as an apoptosis inhibitor and a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups.
  • Abrogates Aβ1-42-induced tau acetylation, memory impairment, and locomotor dysfunction in mice.
  • Potential research area: Alzheimer's disease.
  • Acts as an apoptosis inhibitor.
  • Can be used for research on congenital muscular dystrophy (CMD).
  • Click chemistry reagent with an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups.
Catalog No. 50-003-92572 Supplier Medchemexpress LLC Supplier No. HY16361A50MG
Omigapil maleate (CGP3466B) is an orally active GAPDH nitrosylation inhibitor that can abrogate Aβ1-42-induced tau acetylation, memory impairment, and locomotor dysfunction in mice. It has potential for research into Alzheimer's disease and congenital muscular dystrophy (CMD). Omigapil maleate also functions as an apoptosis inhibitor and a click chemistry reagent, containing an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups.
  • Abrogates Aβ1-42-induced tau acetylation, memory impairment, and locomotor dysfunction in mice.
  • Potential research area: Alzheimer's disease.
  • Acts as an apoptosis inhibitor.
  • Can be used for research on congenital muscular dystrophy (CMD).
  • Click chemistry reagent with an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups.