Learn More
Medchemexpress LLC Omigapil maleate | 200189-97-5 | MFCD09970490 | 99.8% | 391.42 g/mol | C23H21NO5 | 10 MG
SDP
Catalog No. 5000288736
Encompass_Preferred
Omigapil maleate is the maleate salt of omigapil, a small-molecule GAPDH nitrosylation inhibitor and apoptosis modulator used in preclinical research, including studies of congenital muscular dystrophy and neuroprotection. The compound is supplied as a solid of high listed purity and has CAS number 200189-97-5.
  • GAPDH nitrosylation inhibitor and apoptosis modulator.
  • Used in preclinical research for congenital muscular dystrophy and neuroprotection studies.
  • High listed purity suitable for research applications.
  • Available in small pack sizes, including 10 mg, and as a 10 mM solution in DMSO.
  • Store sealed at 4°C away from moisture; in solution store at -80°C for long-term storage.
Catalog No. 50-002-88736 Supplier Medchemexpress LLC Supplier No. HY16361A10MG
Omigapil maleate is the maleate salt of omigapil, a small-molecule GAPDH nitrosylation inhibitor and apoptosis modulator used in preclinical research, including studies of congenital muscular dystrophy and neuroprotection. The compound is supplied as a solid of high listed purity and has CAS number 200189-97-5.
  • GAPDH nitrosylation inhibitor and apoptosis modulator.
  • Used in preclinical research for congenital muscular dystrophy and neuroprotection studies.
  • High listed purity suitable for research applications.
  • Available in small pack sizes, including 10 mg, and as a 10 mM solution in DMSO.
  • Store sealed at 4°C away from moisture; in solution store at -80°C for long-term storage.