Learn More
Selleck Chemical LLC Omipalisib (GSK2126458) 10mM/1mL 1086062-66-9 GSK458
SDP
Supplier:  Selleck Chemical LLC S265810MM/1ML
Encompass_Preferred
Omipalisib (GSK2126458, GSK458) is a highly selective and potent inhibitor of p110?/?/?/?, mTORC1/2 with Ki of 0.019 nM/0.13 nM/0.024 nM/0.06 nM and 0.18 nM/0.3 nM in cell-free assays, respectively. Omipalisib induces autophagy. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6192