Learn More
Description
Blade assemblies for mixing and rotor-stator generator probes for homogenization offer flexibility for blending, suspensions, dispersions, disintegrating, and emulsifications. Speed ranges of 500 to 20,000rpm allow for gentle mixing or higher speed homogenization type applications.
- Quiet operation
- Process sample volumes ranging from 0.30mL to 10L
- Speeds ranging from 500 to 20,000rpm
- Efficiently process most samples in 30 seconds or less
- Interchangeable blade and rotor stator generators for processing within the same container
- Configures to a wide variety of processing attachments including generator probes, cutting blades, and sealed chamber assemblies
- Sealed chamber accessories for aerosol containments
- 600w motor
- Motor stand included
- Processing Range: 0.25mL to 10L
- Power Rating: 600w
- Speed Control: Variable from 500 to 20,000rpm
- On/Off Switch: For easy repeatability
- Dimensions: Height (with stand): 24 in. (60.96cm), Width: 10 in. (25.4cm)
- Weight: 21 lb. (9.5kg)
- Sound Level: <68db
- Soft and Hard Tissue Homogenization
- Cellular Research
- Biological Research
- Food and Plant Analysis
- DNA, RNA, PCR and Protein Extraction
- Enzyme Studies
- Cancer Research
- Pharmaceutical Research
- Animal Tissue
- Medical Research
- Neuroscience
- Emulsions and Suspensions
- Molecular Biology
Compliance
CE approved
Warranty and Services
Three year motor warranty
Specifications
Specifications
| Speed | 500 to 18,000 rpm |
| Voltage | 115 V |
| Wattage | 600 W |
| Capacity (Metric) | 10 L |
| Volume Range (English) | 0.008 to 338.14 oz. |
| Volume Range (Metric) | 10 mL to 10 L |
| Height (English) | 24 in. |
| Height (Metric) | 60.96 cm |
| For Use With (Application) | Ideal for soft and hard tissue homogenization, cellular research, biological research, food and plant analysis, DNA, RNA, PCR and protein extraction, enzyme studies, cancer research, pharmaceutical research, animal tissue, medical research, neuroscience |
| Packaging Quantity | 1 Ea. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
