Learn More
DNA POLYMERASE TECHNOLOGY INC OMNI KLENTAQ 125UL
Catalog No. NC2011916
Encompass
This multipurpose mutant of Klentaq is able to overcome inhibitors in blood, serum, soil, inhibitory foods, and fluorescent dyes. Omni Klentaq can tolerate 20 - 25% whole blood or serum in the PCR reaction.
Catalog No. NC2011916 Supplier DNA POLYMERASE TECHNOLOGY INC Supplier No. 340
This multipurpose mutant of Klentaq is able to overcome inhibitors in blood, serum, soil, inhibitory foods, and fluorescent dyes. Omni Klentaq can tolerate 20 - 25% whole blood or serum in the PCR reaction.