Learn More
MSC Krylon Omni-Pak Can, 16 fl. oz., Flat Latex Blend
Catalog No. 502838344
Encompass
This Omni-Pak can is designed for specialized applications requiring a flat latex blend, delivered as a convenient spray. It features a 16 fl. oz. capacity, providing an efficient solution for various tasks.
  • Designed for flat latex blend applications
  • Features a 16 fl. oz. capacity
  • Delivers product as a spray
  • Extremely flammable aerosol
  • Pressurized container; do not pierce or burn, even after use
  • Protect from sunlight; do not expose to temperatures exceeding 50 °C/122 °F
  • Store in a well-ventilated, cool, dry, and upright place
  • Keep out of reach of children
Catalog No. 50-283-8344 Supplier MSC Supplier No. 37595139
This Omni-Pak can is designed for specialized applications requiring a flat latex blend, delivered as a convenient spray. It features a 16 fl. oz. capacity, providing an efficient solution for various tasks.
  • Designed for flat latex blend applications
  • Features a 16 fl. oz. capacity
  • Delivers product as a spray
  • Extremely flammable aerosol
  • Pressurized container; do not pierce or burn, even after use
  • Protect from sunlight; do not expose to temperatures exceeding 50 °C/122 °F
  • Store in a well-ventilated, cool, dry, and upright place
  • Keep out of reach of children