Learn More
Description
- Suitable for homogenization where sample size or sample hardness dictates that processing with the 7 mm tip is prohibitive.
- Significantly reduces cross-contamination and is virtually maintenance-free.
- Designed for PCR, RNA, and DNA extraction.
- Compatible with Omni TH, THQ, GLH homogenizers, and all automation equipment.
- Omni Tips offer a cost-effective solution for researchers homogenizing samples where contaminants can affect downstream assays.
- Provides a cost-effective solution for researchers homogenizing samples where contaminants may impact downstream assays.
Specifications
Specifications
| Product Type | Probe |
| Length (Metric) Probe | 12 mm |
| Autoclavable | Autoclavable |
| Blade Material | Polypropylene |
| Closure Color | Blue |
| Packaging Quantity | 25/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
