Learn More
LGC GENOMICS LLC (Lucigen) OmniCleave Endonuclease, 50,000 UA robust, flexible endonuclease for the removal/digestion of all types of RNA and DNA leaving only di-, tri- and tetranucleotides behind.