Learn More
Omnifit™ Endpieces for Hit™ Columns
Catalog No. 14533139
Change view
Click to view available options
Diameter (Metric) Inner:
6.6 mm
10 mm
15 mm
Endcapped:
Adjustable
Fixed
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner Endcapped
14-533-139 6.6 mm Adjustable
14-533-140 10 mm Adjustable
14-533-141 15 mm Adjustable
14-533-149 6.6 mm Fixed
14-533-150 10 mm Fixed
14-533-151 15 mm Fixed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-533-139 Supplier Oakton Supplier No. 006HAE06
Mix and match Omnifit™ Endpieces for Hit™ Columns for your appliction
Mix and match endpieces to create a column for your application
Specifications
Endcapped Adjustable
Type Liquid Chromatography Column
Diameter (Metric) Inner 6.6 mm
For Use With (Equipment) Omnifit™ HiT Chromatography Columns
Material PTFE