Learn More
Cole-Parmer Diba Omnifit EZ Replacement Glass, 10 x 330 mm; 1/ea
Supplier:  Cole-Parmer 1194236
Encompass_Preferred
You can mix and match column length, while using your existing endpieces for ultimate flexibility. You can use a different length tubes with your existing endpieces to meet specific requirements.
Catalog No. NC1482468