Learn More
MSC Carlisle Mop Bucket Combo, 35 qt, Side Press Wringer, Polypropylene, Blue
Catalog No. 502838744
Encompass
This 35 qt mop bucket combo in blue provides a durable and efficient cleaning solution, featuring a side press wringer and a soiled water insert. Constructed from corrosion-resistant polypropylene, it is designed for long-lasting commercial use with a wringer rated for 200,000 cycles. It incorporates anti-splash technology and an anti-tip design to minimize spills and prevent overturning. A key feature is the insert versatility, separating clean and soiled water for hygienic cleaning. Mobility is enhanced by scuff-resistant, non-marking, swiveling casters, providing mess-free transport. An ergonomic handle simplifies movement and wringing.
  • Durable polypropylene construction.
  • 200,000-cycle wringer.
  • Anti-splash technology.
  • Anti-tip design.
  • Soiled water insert.
  • Scuff-resistant swivel casters.
  • Ergonomic handle.
  • Recyclable (Polypropylene 5).
Catalog No. 50-283-8744 Supplier MSC Supplier No. 73924185
This 35 qt mop bucket combo in blue provides a durable and efficient cleaning solution, featuring a side press wringer and a soiled water insert. Constructed from corrosion-resistant polypropylene, it is designed for long-lasting commercial use with a wringer rated for 200,000 cycles. It incorporates anti-splash technology and an anti-tip design to minimize spills and prevent overturning. A key feature is the insert versatility, separating clean and soiled water for hygienic cleaning. Mobility is enhanced by scuff-resistant, non-marking, swiveling casters, providing mess-free transport. An ergonomic handle simplifies movement and wringing.
  • Durable polypropylene construction.
  • 200,000-cycle wringer.
  • Anti-splash technology.
  • Anti-tip design.
  • Soiled water insert.
  • Scuff-resistant swivel casters.
  • Ergonomic handle.
  • Recyclable (Polypropylene 5).