Learn More
Omnimed Inc OmniGuard With Audit Lock (15" H x 14" W x 6" D)
SDP
Catalog No. NC3956413
Encompass_Preferred
Introducing our latest Narcotic Cabinet, designed with security and convenience in mind. It features a secure electronic lock, complemented by a manager's key for easy lockout bypass. The cabinet supports up to 20 customizable user codes, offering flexibility and enhanced protection for controlled substances. With compact dimensions of 15" H x 14" W x 6" D, it's the ideal solution for secure storage.
Catalog No. NC3956413 Supplier Omnimed Inc Supplier No. 291666
Introducing our latest Narcotic Cabinet, designed with security and convenience in mind. It features a secure electronic lock, complemented by a manager's key for easy lockout bypass. The cabinet supports up to 20 customizable user codes, offering flexibility and enhanced protection for controlled substances. With compact dimensions of 15" H x 14" W x 6" D, it's the ideal solution for secure storage.