Learn More
Omnimed Inc Omnimed Acrylic Draft Shield, Medium, 15"W x 16"D x 15"H
SDP
Catalog No. NC3956525
Encompass_Preferred
Omnimed's Acrylic Draft Shield is a simple addition to your lab workstation that values space occupancy and the need for accurate readings. Ideal for laboratories and science classrooms where delicate equipment demands precise assessment. The protection shield is transparent, lightweight, easy-to-clean, and features a cut-out in the back for neat cable management. Compact design and slim frame of the draft enclosure provides minimal space occupancy across any tabletop or desk to ensure simple and fast operation of your lab scale.
Catalog No. NC3956525 Supplier Omnimed Inc Supplier No. 307211
Omnimed's Acrylic Draft Shield is a simple addition to your lab workstation that values space occupancy and the need for accurate readings. Ideal for laboratories and science classrooms where delicate equipment demands precise assessment. The protection shield is transparent, lightweight, easy-to-clean, and features a cut-out in the back for neat cable management. Compact design and slim frame of the draft enclosure provides minimal space occupancy across any tabletop or desk to ensure simple and fast operation of your lab scale.