Learn More
Omnimed Inc Omnimed Triple Bulk Dispenser, Clear Front
SDP
Supplier:  Omnimed Inc 307033
Encompass_Preferred
Introducing Omnimed's Bulk Dispenser -Triple Bin, a versatile and reliable solution designed to streamline storage and dispensing of protective apparel and miscellaneous items. Crafted with precision from high-quality Stainless Steel and PETG, this dispenser is built to withstand the demands of various environments, Ideal for use in surgical areas, interventional radiology, clean rooms, and beyond. Upgrade your storage capabilities and enhance workflow efficiency with Omnimed's commitment to quality and innovation. The new composite bulk dispensers fuse two materials together to create a superior product to any dispensers currently on the market. The stainless steel shell provides strength and longevity while the PTEG high impact plastic provides aesthetics and visualization.
Catalog No. NC3956506