Learn More
Enzo Life Sciences OMNIMMP fluorogenic substrate (1mg)
SDP
Catalog No. 502011240
Encompass_Preferred
Fluorogenic substrate for nearly all matrix metalloproteinase (MMP) enzymes, and ADAM17/TACE. This highly-quenched (99.5%) substrate is useful for inhibitor screening and kinetic analysis. Ex.: 328 nm, Em.: 393 nm, although the following Ex/Em have been used: 320-340/400-420. Also available: OMNIMMP Fluorogenic Control Peptide (BML-P127). Alternative name: Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH2 . AcOH, Matrix metalloproteinase substrate. Purity: ≥98%. Solubility: Soluble in methanol (1mg/ml) or DMSO (20mg/ml). Long Term Storage: -20°C. Applications: WB Assay.
Catalog No. 50-201-1240 Supplier Enzo Life Sciences Supplier No. BMLP1260001
Fluorogenic substrate for nearly all matrix metalloproteinase (MMP) enzymes, and ADAM17/TACE. This highly-quenched (99.5%) substrate is useful for inhibitor screening and kinetic analysis. Ex.: 328 nm, Em.: 393 nm, although the following Ex/Em have been used: 320-340/400-420. Also available: OMNIMMP Fluorogenic Control Peptide (BML-P127). Alternative name: Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH2 . AcOH, Matrix metalloproteinase substrate. Purity: ≥98%. Solubility: Soluble in methanol (1mg/ml) or DMSO (20mg/ml). Long Term Storage: -20°C. Applications: WB Assay.