Learn More
Enzo Life Sciences OMNIMMP RED fluorogenic substrate (100µg)
SDP
Catalog No. 502011330
Encompass_Preferred
Highly-quenched, ultra-bright fluorogenic substrate for most MMPs. 6’-TAMRA fluorescence is thoroughly quenched by the TQ3 group until cleavage by MMPs separates the two moieties. The OMNIMMP RED substrate offers key advantages over other MMP substrates. Purity: ≥90% (HPLC). Solubility: Soluble in DMSO (50mM). Long Term Storage: -20°C.
Catalog No. 50-201-1330 Supplier Enzo Life Sciences Supplier No. BMLP2770100
Highly-quenched, ultra-bright fluorogenic substrate for most MMPs. 6’-TAMRA fluorescence is thoroughly quenched by the TQ3 group until cleavage by MMPs separates the two moieties. The OMNIMMP RED substrate offers key advantages over other MMP substrates. Purity: ≥90% (HPLC). Solubility: Soluble in DMSO (50mM). Long Term Storage: -20°C.