Learn More
Electron Microscopy Sciences Omnimount™ solvent, 100ml, Compatible with HistoClear II
SDP
Catalog No. 501900432
Encompass_Preferred
Specially developed mounting medium is compatible with National Diagnostic clearing agents, HistoClear II. Omnimount combines outstanding optical characteristics with low fluororescence and exceptional durability. Omnimount is a truly universal mounting medium compatible with all common clearing agents: xylene, toluene, limonene, and petroleum derived products.
The Omnimount solvent has a higher flash point and a lower toxicity than xylene based mountants, so Omnimount provides both a safer work environment and reduce shipping costs.
Catalog No. 50-190-0432 Supplier Electron Microscopy Sciences Supplier No. 1799701
Specially developed mounting medium is compatible with National Diagnostic clearing agents, HistoClear II. Omnimount combines outstanding optical characteristics with low fluororescence and exceptional durability. Omnimount is a truly universal mounting medium compatible with all common clearing agents: xylene, toluene, limonene, and petroleum derived products.
The Omnimount solvent has a higher flash point and a lower toxicity than xylene based mountants, so Omnimount provides both a safer work environment and reduce shipping costs.