Learn More
National Diagnostics OMNIMOUNT 100 ML
SDP
Catalog No. 5089990146
Encompass
Omnimount, Universally compatible synthetic mounting medium. Ideally suited for use with HistoClear II, 100 ML
Catalog No. 50-899-90146 Supplier National Diagnostics Supplier No. HS110100ML
Omnimount, Universally compatible synthetic mounting medium. Ideally suited for use with HistoClear II, 100 ML