Learn More
Med Vet International Omnipaque Iohexol Injection 240mg/mL, 50mL
Catalog No. 502827863
Encompass_Preferred
Omnipaque (iohexol injection) is a radiographic contrast medium indicated for intrathecal administration in adults including myelography (lumbar, thoracic, cervical, total columnar) and in contrast enhancement for computerized tomography (myelography, cisternography, ventriculography).
Catalog No. 50-282-7863 Supplier Med Vet International Supplier No. RXOMNIPAQUE240MG50ML
Omnipaque (iohexol injection) is a radiographic contrast medium indicated for intrathecal administration in adults including myelography (lumbar, thoracic, cervical, total columnar) and in contrast enhancement for computerized tomography (myelography, cisternography, ventriculography).