Learn More
Alkali Scientific OmniPette™ Single Channel Pipetter1 to 5mL, No Ejector Button on this Model
SDP
Catalog No. 502335834
Encompass_Preferred
The OmniPette™ series of single channel micro pipetters is a culmination of decades of liquid handling research. Every aspect of accuracy, reliability, and user comfort has been carefully reviewed in designing these high precision pipettes. The result is an advanced pipette that exhibits superior accuracy & ergonomics and is comfortable to use, even during prolonged and repetitive pipetting. The OmniPette™ series features a continuously adjustable, digital micrometer for volume selection and a non-mandatory locking wheel (positioned at the top of the handle for simple one-handed activation/deactivation). The finely polished stainless piston and chemically resistant, maintenance-free seals provide unsurpassed accuracy. The air-jacketed handle protects the micrometer mechanism from the inaccuracies that can be caused by thermal expansion. This enhances continued pipetting precision even after prolonged use.
Catalog No. 50-233-5834 Supplier Alkali Scientific Supplier No. P92805M
The OmniPette™ series of single channel micro pipetters is a culmination of decades of liquid handling research. Every aspect of accuracy, reliability, and user comfort has been carefully reviewed in designing these high precision pipettes. The result is an advanced pipette that exhibits superior accuracy & ergonomics and is comfortable to use, even during prolonged and repetitive pipetting. The OmniPette™ series features a continuously adjustable, digital micrometer for volume selection and a non-mandatory locking wheel (positioned at the top of the handle for simple one-handed activation/deactivation). The finely polished stainless piston and chemically resistant, maintenance-free seals provide unsurpassed accuracy. The air-jacketed handle protects the micrometer mechanism from the inaccuracies that can be caused by thermal expansion. This enhances continued pipetting precision even after prolonged use.