Learn More
Alkali Scientific OmniPette™ Digital Pipette Controller
SDP
Catalog No. 5000662992
Encompass_Preferred
OmniPette™ Digital Pipette Controllers add convenience and functionality to the OmniPette™ line, allowing users to easily control aspirate and dispense speeds during pipetting tasks. Integrated buttons and a clear digital display enable quick adjustment of maximum aspirate and dispense rates while working. Pressure sensitive triggers provide smooth and precise flow control for improved accuracy. The display also monitors battery level, and the unit charges through a convenient USB C port. Designed for comfort, the light controller (6.3 oz / 170 g) fits comfortably in either hand and features an ergonomic body with balanced distribution. The OmniPette™ controller also includes the proprietary Quickstand™ retractable stand, allowing the instrument to be safely placed on the bench without contamination.
Catalog No. 50-006-62992 Supplier Alkali Scientific Supplier No. P7290
OmniPette™ Digital Pipette Controllers add convenience and functionality to the OmniPette™ line, allowing users to easily control aspirate and dispense speeds during pipetting tasks. Integrated buttons and a clear digital display enable quick adjustment of maximum aspirate and dispense rates while working. Pressure sensitive triggers provide smooth and precise flow control for improved accuracy. The display also monitors battery level, and the unit charges through a convenient USB C port. Designed for comfort, the light controller (6.3 oz / 170 g) fits comfortably in either hand and features an ergonomic body with balanced distribution. The OmniPette™ controller also includes the proprietary Quickstand™ retractable stand, allowing the instrument to be safely placed on the bench without contamination.