Learn More
Alkali Scientific OmniPette Single Channel Pipetter 5 to 50 ul
SDP
Catalog No. 502389531
Encompass_Preferred
The OmniPette™ series of single channel micro pipetters is a culmination of decades of liquid handling research. Every aspect of accuracy, reliability, and user comfort has been carefully reviewed in designing these high precision pipettes. The result is an advanced pipette that exhibits superior accuracy & ergonomics and is comfortable to use, even during prolonged and repetitive pipetting.Reduced actuation forcesStainless steel pistons and teflon sealsSuperior accuracy and reproducibilityAdjustable Volume & Locking WheelFull Volume Range (0.1µL - 10mL)Outstanding User ComfortFully autoclavable3 year warranty
Catalog No. 50-238-9531 Supplier Alkali Scientific Supplier No. P928050U
The OmniPette™ series of single channel micro pipetters is a culmination of decades of liquid handling research. Every aspect of accuracy, reliability, and user comfort has been carefully reviewed in designing these high precision pipettes. The result is an advanced pipette that exhibits superior accuracy & ergonomics and is comfortable to use, even during prolonged and repetitive pipetting.Reduced actuation forcesStainless steel pistons and teflon sealsSuperior accuracy and reproducibilityAdjustable Volume & Locking WheelFull Volume Range (0.1µL - 10mL)Outstanding User ComfortFully autoclavable3 year warranty