Learn More
MilliporeSigma Omnipore Membrane Filter, 5.0 um, 47 mm diameter
Catalog No. NC1803996
Encompass
Hydrophobic, polytetrafluoroethylene (PTFE) polymer membrane bonded to a high density polyethylene support to improve the handling characteristics of the filter for normal applications. Fluoropore membranes provide broad chemical compatibility, high flow rates and consistency.
Catalog No. NC1803996 Supplier MilliporeSigma Supplier No. JMWP04700
Add to Cart
Add to Cart
Hydrophobic, polytetrafluoroethylene (PTFE) polymer membrane bonded to a high density polyethylene support to improve the handling characteristics of the filter for normal applications. Fluoropore membranes provide broad chemical compatibility, high flow rates and consistency.